ADVH49B8354F AChemBlock

1,1'-di-tert-butyl-4'-hydroxy-[3,4'-bipiperidin]-4-one

  • Cas Number : 2101206-63-5
  • Molecular Weight : 310.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-1-di-tert-butyl-4-hydroxy-3-4-bipiperidin-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88776
Availability details for
Quantity
  • Availability
  • Add to Cart