ADVH49B83552 AChemBlock

tert-butyl 4-((trimethylsilyl)oxy)-3,6-dihydropyridine-1(2H)-carboxylate

  • Cas Number : 211108-48-4
  • Molecular Weight : 271.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-trimethylsilyl-oxy-3-6-dihydropyridine-1-2h-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88779
Availability details for
Quantity
  • Availability
  • Add to Cart