ADVH49B83553 AChemBlock

1-(1-(4-(chloromethyl)phenyl)ethoxy)-2,2,6,6-tetramethylpiperidine

  • Cas Number : 212132-38-2
  • Molecular Weight : 309.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-1-4-chloromethyl-phenyl-ethoxy-2-2-6-6-tetramethylpiperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88780
Availability details for
Quantity
  • Availability
  • Add to Cart