ADVH49B83554 AChemBlock

ethyl (Z)-3-(pyridin-4-yl)acrylate

  • Cas Number : 24489-96-1
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-z-3-pyridin-4-yl-acrylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88781
Availability details for
Quantity
  • Availability
  • Add to Cart