ADVH49B83555 AChemBlock

1-(5-ethyl-2-hydroxyphenyl)ethan-1-one

  • Cas Number : 24539-92-2
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-5-ethyl-2-hydroxyphenyl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88782
Availability details for
Quantity
  • Availability
  • Add to Cart