ADVH49B83558 AChemBlock

2-chloro-N-methyl-N-(4-nitrophenyl)acetamide

  • Cas Number : 2653-16-9
  • Molecular Weight : 228.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-n-methyl-n-4-nitrophenyl-acetamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88787
Availability details for
Quantity
  • Availability
  • Add to Cart