ADVH49B83562 AChemBlock

manganese(III) 2,2'-((1E,1'E)-(ethane-1,2-diylbis(azaneylylidene))bis(methaneylylidene))diphenolate chloride

  • Cas Number : 53177-12-1
  • Molecular Weight : 356.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/manganese-iii-2-2-1e-1-e-ethane-1-2-diylbis-azaneylylidene-bis-methaneylylidene-diphenolate-chloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88800
Availability details for
Quantity
  • Availability
  • Add to Cart