ADVH49B83564 AChemBlock

2,2-difluoropent-4-enoic acid

  • Cas Number : 55039-89-9
  • Molecular Weight : 136.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-difluoropent-4-enoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88802
Availability details for
Quantity
  • Availability
  • Add to Cart