ADVH49B83565 AChemBlock

tert-butyl (3-chloro-2-hydroxypropyl)carbamate

  • Cas Number : 570390-94-2
  • Molecular Weight : 209.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-chloro-2-hydroxypropyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88803
Availability details for
Quantity
  • Availability
  • Add to Cart