ADVH49B83566 AChemBlock

tert-butyl 4-fluoro-4-formylpiperidine-1-carboxylate

  • Cas Number : 614731-09-8
  • Molecular Weight : 231.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-fluoro-4-formylpiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88804
Availability details for
Quantity
  • Availability
  • Add to Cart