ADVH49B8356A AChemBlock

tert-butyl 5-fluoro-4-phenyl-3,6-dihydropyridine-1(2H)-carboxylate

  • Cas Number : 651321-26-5
  • Molecular Weight : 277.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-fluoro-4-phenyl-3-6-dihydropyridine-1-2h-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88808
Availability details for
Quantity
  • Availability
  • Add to Cart