ADVH49B8356C AChemBlock

2,2,6,6-tetramethylpiperidin-1-ol

  • Cas Number : 7031-93-8
  • Molecular Weight : 157.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-6-6-tetramethylpiperidin-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88810
Availability details for
Quantity
  • Availability
  • Add to Cart