ADVH49B83571 AChemBlock

naphthalen-2-yl 4-methylbenzenesulfonate

  • Cas Number : 7385-85-5
  • Molecular Weight : 298.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/naphthalen-2-yl-4-methylbenzenesulfonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88816
Availability details for
Quantity
  • Availability
  • Add to Cart