ADVH49B83573 AChemBlock

tert-butyl (S)-(3-(3,4-dihydroxyphenyl)-1-(dimethylamino)-1-oxopropan-2-yl)carbamate

  • Cas Number : 752251-02-8
  • Molecular Weight : 324.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-s-3-3-4-dihydroxyphenyl-1-dimethylamino-1-oxopropan-2-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88818
Availability details for
Quantity
  • Availability
  • Add to Cart