ADVH49B83574 AChemBlock

8-benzyl-8-azabicyclo[3.2.1]octan-3-amine

  • Cas Number : 76272-35-0
  • Molecular Weight : 216.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-benzyl-8-azabicyclo-3-2-1-octan-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88819
Availability details for
Quantity
  • Availability
  • Add to Cart