ADVH49B83577 AChemBlock

adamantan-1-ylmethyl acetate

  • Cas Number : 778-11-0
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/adamantan-1-ylmethyl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88822
Availability details for
Quantity
  • Availability
  • Add to Cart