ADVH49B83579 AChemBlock

ethyl 1-benzyl-3-hydroxypiperidine-4-carboxylate

  • Cas Number : 850040-08-3
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-benzyl-3-hydroxypiperidine-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88824
Availability details for
Quantity
  • Availability
  • Add to Cart