ADVH49B8357A AChemBlock

1'-benzyl-7-methoxyspiro[chromane-2,4'-piperidin]-4-one

  • Cas Number : 868361-89-1
  • Molecular Weight : 337.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-7-methoxyspiro-chromane-2-4-piperidin-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88825
Availability details for
Quantity
  • Availability
  • Add to Cart