ADVH49B8357B AChemBlock

ethyl 3-(2,2-dimethyl-4,6-dioxo-1,3-dioxan-5-yl)propanoate

  • Cas Number : 90734-80-8
  • Molecular Weight : 244.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-2-2-dimethyl-4-6-dioxo-1-3-dioxan-5-yl-propanoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88828
Availability details for
Quantity
  • Availability
  • Add to Cart