ADVH49B8357C AChemBlock

tert-butyl 4-(4-methylbenzoyl)piperidine-1-carboxylate

  • Cas Number : 912768-78-6
  • Molecular Weight : 303.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-4-methylbenzoyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88829
Availability details for
Quantity
  • Availability
  • Add to Cart