ADVH49B83580 AChemBlock

dimethyl (2S,4R)-piperidine-2,4-dicarboxylate

  • Cas Number : 98935-65-0
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/dimethyl-2s-4r-piperidine-2-4-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88836
Availability details for
Quantity
  • Availability
  • Add to Cart