ADVH49B83581 AChemBlock

(E)-N-(2,4-dimethylphenyl)-2,2,2-trifluoro-N'-((3,5,6-trimethylpyrazin-2-yl)methylene)acetohydrazide

  • Molecular Weight : 364.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/e-n-2-4-dimethylphenyl-2-2-2-trifluoro-n-3-5-6-trimethylpyrazin-2-yl-methylene-acetohydrazide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88837
Availability details for
Quantity
  • Availability
  • Add to Cart