ADVH49B83583 AChemBlock

methyl spiro[adamantane-2,2'-[1,3]dioxolane]-5-carboxylate

  • Molecular Weight : 252.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-spiro-adamantane-2-2-1-3-dioxolane-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88839
Availability details for
Quantity
  • Availability
  • Add to Cart