ADVH49B83584 AChemBlock

benzyl 3,3-difluoro-4-methylenepiperidine-1-carboxylate

  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/benzyl-3-3-difluoro-4-methylenepiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88840
Availability details for
Quantity
  • Availability
  • Add to Cart