ADVH49B83586 AChemBlock

8-bromo-2,2-dimethyl-6-(methylsulfonyl)-2H-benzo[b][1,4]oxazin-3(4H)-one

  • Molecular Weight : 334.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-bromo-2-2-dimethyl-6-methylsulfonyl-2h-benzo-b-1-4-oxazin-3-4h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88637
Availability details for
Quantity
  • Availability
  • Add to Cart