ADVH49B83589 AChemBlock

(S)-3,3-difluoro-1-methylpiperidin-4-ol

  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-3-difluoro-1-methylpiperidin-4-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88845
Availability details for
Quantity
  • Availability
  • Add to Cart