ADVH49B8358A AChemBlock

(S)-3,3-difluoropiperidin-4-ol hydrochloride

  • Molecular Weight : 173.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-3-difluoropiperidin-4-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88846
Availability details for
Quantity
  • Availability
  • Add to Cart