ADVH49B8358C AChemBlock

3,3-difluoro-1-propylpiperidin-4-ol hydrochloride

  • Molecular Weight : 215.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-3-difluoro-1-propylpiperidin-4-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88848
Availability details for
Quantity
  • Availability
  • Add to Cart