ADVH49B8358E AChemBlock

1-benzyl-3,3-difluoropiperidin-4-one hydrate

  • Molecular Weight : 243.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-3-3-difluoropiperidin-4-one-hydrate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88850
Availability details for
Quantity
  • Availability
  • Add to Cart