ADVH49B8358F AChemBlock

benzyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate

  • Molecular Weight : 287.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/benzyl-3-3-difluoro-4-oxopiperidine-1-carboxylate-hydrate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88851
Availability details for
Quantity
  • Availability
  • Add to Cart