ADVH49B83592 AChemBlock

methyl 2-(1-(cyclopropylmethyl)-1H-indol-2-yl)-7-methoxy-1-methyl-1H-benzo[d]imidazole-5-carboxylate

  • Molecular Weight : 389.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-1-cyclopropylmethyl-1h-indol-2-yl-7-methoxy-1-methyl-1h-benzo-d-imidazole-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88640
Availability details for
Quantity
  • Availability
  • Add to Cart