ADVH49B83599 AChemBlock

ethyl 2-(3-hydroxy-4-oxocyclohexyl)acetate

  • Molecular Weight : 200.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-3-hydroxy-4-oxocyclohexyl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88861
Availability details for
Quantity
  • Availability
  • Add to Cart