ADVH49B8359B AChemBlock

tert-butyl 2-methyl-1-oxa-7-azaspiro[3.5]nonane-7-carboxylate

  • Molecular Weight : 241.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-methyl-1-oxa-7-azaspiro-3-5-nonane-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88863
Availability details for
Quantity
  • Availability
  • Add to Cart