ADVH49B8359C AChemBlock

1,2-dimethoxyanthracene-9,10-dione

  • Molecular Weight : 268.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-dimethoxyanthracene-9-10-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88864
Availability details for
Quantity
  • Availability
  • Add to Cart