ADVH49B8359E AChemBlock

(S)-2-amino-3-(4-(trifluoromethyl)phenyl)propanoic acid

  • Cas Number : 114872-99-0
  • Molecular Weight : 233.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-3-4-trifluoromethyl-phenyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88868
Availability details for
Quantity
  • Availability
  • Add to Cart