ADVH49B835A0 AChemBlock

(R)-3-amino-3-(4-chlorophenyl)propanoic acid

  • Cas Number : 131690-61-4
  • Molecular Weight : 199.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-amino-3-4-chlorophenyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88878
Availability details for
Quantity
  • Availability
  • Add to Cart