ADVH49B835A1 AChemBlock

(R)-3-amino-3-phenylpropanoic acid

  • Cas Number : 13921-90-9
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-amino-3-phenylpropanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88880
Availability details for
Quantity
  • Availability
  • Add to Cart