ADVH49B835A5 AChemBlock

3-(2,6-difluoropyridin-4-yl)-4-methylaniline

  • Molecular Weight : 220.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-6-difluoropyridin-4-yl-4-methylaniline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88642
Availability details for
Quantity
  • Availability
  • Add to Cart