ADVH49B835BF AChemBlock

3-(aminomethyl)-4-chloro-7-(2-hydroxyethoxy)benzo[c][1,2]oxaborol-1(3H)-ol

  • Molecular Weight : 257.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-aminomethyl-4-chloro-7-2-hydroxyethoxy-benzo-c-1-2-oxaborol-1-3h-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88644
Availability details for
Quantity
  • Availability
  • Add to Cart