ADVH49B835C2 AChemBlock

(S)-morpholin-3-ylmethanol

  • Cas Number : 211053-50-8
  • Molecular Weight : 117.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-morpholin-3-ylmethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88226
Availability details for
Quantity
  • Availability
  • Add to Cart