ADVH49B835C6 AChemBlock

2-((tert-butoxycarbonyl)amino)heptanoic acid

  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-tert-butoxycarbonyl-amino-heptanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89112
Availability details for
Quantity
  • Availability
  • Add to Cart