ADVH49B835CB AChemBlock

(S)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(4-methoxyphenyl)butanoic acid

  • Molecular Weight : 431.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-9h-fluoren-9-yl-methoxy-carbonyl-amino-4-4-methoxyphenyl-butanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89117
Availability details for
Quantity
  • Availability
  • Add to Cart