ADVH49B835D3 AChemBlock

3-((tert-butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid

  • Molecular Weight : 310.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-tert-butoxycarbonyl-amino-3-4-nitrophenyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89125
Availability details for
Quantity
  • Availability
  • Add to Cart