ADVH49B835DD AChemBlock

methyl (R)-2-(2-chlorophenyl)-2-(((4-nitrophenyl)sulfonyl)oxy)acetate

  • Molecular Weight : 385.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-r-2-2-chlorophenyl-2-4-nitrophenyl-sulfonyl-oxy-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88650
Availability details for
Quantity
  • Availability
  • Add to Cart