ADVH49B835DE AChemBlock

2-methyl-1,2,3,4-tetrahydroisoquinolin-6-ol

  • Cas Number : 14097-39-3
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-1-2-3-4-tetrahydroisoquinolin-6-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89136
Availability details for
Quantity
  • Availability
  • Add to Cart