ADVH49B835DF AChemBlock

4-amino-1H-pyrrole-2-carboxylic acid

  • Cas Number : 155815-95-5
  • Molecular Weight : 126.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-amino-1h-pyrrole-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89137
Availability details for
Quantity
  • Availability
  • Add to Cart