ADVH49B835E1 AChemBlock

2-amino-3-bromo-4-chlorobenzoic acid

  • Cas Number : 1698146-49-4
  • Molecular Weight : 250.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-3-bromo-4-chlorobenzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89139
Availability details for
Quantity
  • Availability
  • Add to Cart