ADVH49B835E2 AChemBlock

3-(2-bromo-4-fluorophenyl)propanoic acid

  • Cas Number : 174603-55-5
  • Molecular Weight : 247.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-2-bromo-4-fluorophenyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89140
Availability details for
Quantity
  • Availability
  • Add to Cart