ADVH49B835E6 AChemBlock

4-hydroxyisoquinolin-1(2H)-one

  • Cas Number : 30081-72-2
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-hydroxyisoquinolin-1-2h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89144
Availability details for
Quantity
  • Availability
  • Add to Cart