ADVH49B835EB AChemBlock

5-(tert-butyl)-1H-pyrrole-2-carbaldehyde

  • Cas Number : 59001-10-4
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-tert-butyl-1h-pyrrole-2-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89149
Availability details for
Quantity
  • Availability
  • Add to Cart